missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, 150,000 μS/cm (150 mS/cm) at 25°C, Ricca Chemical

Catalog No. 22491532
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22491532 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22491532 Supplier Ricca Chemical Company Supplier No. 2249.1532

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Conductivity Standard
Packaging Natural Poly Bottle
CAS 7647-14-5, 7732-18-5
CAS Min % 89.0, 8.62
CAS Max % 93.0, 8.98
Physical Form Liquid
Color Colorless
pH 6.7 to 7.3
Boiling Point Approximately 100.1°C
For Use With (Equipment) Conductivity Meters and Probes
Solubility Miscible with water
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 1 L
Grade Laboratory
Show More Show Less