missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard, 15 μS/cm at 25°C, Ricca Chemical
Spécifications
Spécifications
| Chemical Name or Material | Conductivity Standard |
| Packaging | Natural Poly Bottle |
| Name Note | 15μS/cm (15μmho/cm) at 25°C |
| CAS | 7647-14-5, 7732-18-5 |
| CAS Min % | 98.0, 7.0E-4 |
| CAS Max % | 100.0, 7.0E-4 |
| Physical Form | Liquid |
| Color | Colorless |
| Boiling Point | 100°C |
| Solution Type | Conductivity Standard |
| Afficher plus de résultats |