missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, 15 μS/cm at 25°C, Ricca Chemical

Catalog No. 22360316
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22360316 500 mL
R22360301A 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22360316 Supplier Ricca Chemical Company Supplier No. 2236.0316
Only null left

Specifications

Chemical Name or Material Conductivity Standard
Packaging Natural Poly Bottle
Name Note 15μS/cm (15μmho/cm) at 25°C
CAS 7647-14-5, 7732-18-5
CAS Min % 98.0, 7.0E-4
CAS Max % 100.0, 7.0E-4
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 500 mL
Show More Show Less