missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard - 147 mhos/cm, MilliporeSigma™ Supelco™

Catalog No. p-7187480
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111004895 500 mL
111004690 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111004895 Supplier Merck Emd Millipore Supplier No. COND147500ML
Only null left

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Featured Industries: Environmental

Specifications

Type Certified Reference Material
Quantity 500 mL
Grade Certified Reference Material