missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, 1413 μS/cm at 25°C NIST Traceable, Reagents

Catalog No. 01336679
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336679 500 mL
01336682 1 L
01336681 20 L
01336680 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01336679 Supplier Labchem Inc Supplier No. C7001413500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Conductivity Standard, 1413 μS/cm at 25°C NIST Traceable, Standard, use for Conductivity meter calibration. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.

Specifications

Chemical Name or Material Conductivity Standard, 1413 μS/cm at 25°C NIST Traceable
Packaging Poly Bottle
CAS 7732-18-5, 7647-14-5
Specific Gravity 0.9986
Physical Form Liquid
Molecular Formula NaCl/H2O
Technique Calibration of Conductivity Instrument
Color Colorless
pH 6.7 to 7.3
Boiling Point 100°C
For Use With (Equipment) Conductivity Meter
Solubility Completely miscible
Type Conductivity Standard
Quantity 500 mL
Show More Show Less