missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, 1412 μS/cm at 25°C Sodium Chloride, Reagents

Catalog No. 01335445
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335445 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335445 Supplier Labchem Inc Supplier No. CC981412500A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Conductivity Standard, 1412 μS/cm at 25°C Sodium Chloride, NIST Traceable, use for Conductivity meter calibration. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.

Specifications

Chemical Name or Material Conductivity Standard, 1412 μS/cm at 25°C Sodium Chloride
Packaging Poly Bottle
CAS 7732-18-5, 7647-14-5
Specific Gravity 1.0004
Physical Form Liquid
Molecular Formula NaCl/H2O
Technique Calibration of Conductivity Instrument
Color Colorless
pH 6.7 to 7.3
Boiling Point 100°C
For Use With (Equipment) Conductivity Meter
Solubility Completely miscible
Quantity 500 mL
Show More Show Less