missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, 1409μmho/cm, Certified, LabChem™

Catalog No. p-5380886
Change view
Click to view available options
Quantity:
1 L
10 L
Packaging:
Cubitainer™
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC187802 1 L Poly Bottle
LC187806 10 L Cubitainer™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC187802 Supplier LabChem Supplier No. LC187802
Only null left

Specifications

Chemical Name or Material Conductivity Standard, 1409μmho/cm
Name Note 1409μmho/cm
Packaging Poly Bottle
CAS 7447-40-7, 7732-18-5
CAS Max % 99.93, 0.07
Identification Passes Test
Density 1g/mL
Specific Gravity 1g/mL
Conductivity 1409umho/cm ± 20
Physical Form Liquid
Linear Formula KCl
Color Colorless
Solubility Soluble in water
Quantity 1 L
Formula Weight 74.55
Grade Certified
Show More Show Less