missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Conductivity Standard, 12.9 μS/cm at 25°C, Ricca Chemical

$159.70 - $274.03

Specifications

Chemical Name or Material Conductivity Standard
Packaging Natural Poly Bottle
pH Approximately 7
Boiling Point 100°C
Solution Type Conductivity Standard
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
588513-16
View Documents
Ricca Chemical Company
58851316
500 mL
Each for $159.70
Only null left
 
588513-32
View Documents
Ricca Chemical Company
58851332
1 L
Each for $274.03
Only null left
 
Description

Description

Specifications

Specifications

Conductivity Standard
Approximately 7
Conductivity Standard
Potassium Chloride in Water
Natural Poly Bottle
100°C
Conductivity Standard
Videos
SDS
Documents

Documents