missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard, 10 μS/cm at 25°C (4.7 ppm TDS as NaCl), Ricca Chemical
Supplier: Ricca Chemical Company RI223632
Specifications
| Conductivity Standard | |
| Natural Poly Bottle | |
| 98.0,5.0E-4 | |
| Liquid | |
| 6.7 to 7.3 | |
| 100°C | |
| Conductivity Standard | |
| 1 L |
| 10μS/cm (10μmho/cm) at 25°C | |
| 7647-14-5,7732-18-5 | |
| 100.0,5.0E-4 | |
| Colorless | |
| 0°C | |
| Completely miscible | |
| Conductivity Standard | |
| 4.7 ppm as NaCl |