missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard, 10 μS/cm at 25°C (4.7 ppm TDS as NaCl), Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Conductivity Standard |
| Packaging | Pack |
| Name Note | 10μS/cm (10μmho/cm) at 25°C |
| CAS | 7647-14-5, 7732-18-5 |
| CAS Min % | 98.0, 5.0E-4 |
| CAS Max % | 100.0, 5.0E-4 |
| Physical Form | Liquid |
| Color | Colorless |
| pH | 6.7 to 7.3 |
| Melting Point | 0°C |
| Show More |