missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, 10,000 μS/cm (10 mS/cm) at 25°C, Ricca Chemical

Catalog No. 22461016
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22461016 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22461016 Supplier Ricca Chemical Company Supplier No. 2246.1016
Only null left

5400 ppm TDS as NaCl

Specifications

Chemical Name or Material Conductivity Standard
Packaging Natural Poly Bottle
Name Note With attached Certificate of Analysis
CAS 7647-14-5, 7732-18-5
CAS Min % 97.0, 0.53
CAS Max % 100.0, 0.55
Physical Form Liquid
Color Colorless
pH 6.7 to 7.3
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) Conductivity Meters and Probes
Solubility Miscible with water
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 500 mL
Grade Laboratory
Show More Show Less