missing translation for 'onlineSavingsMsg'
Learn More

Conductivity Standard, 1413 μS/cm at 25°C, Ricca Chemical

Catalog No. 22365232
Change view
Click to view available options
Quantity:
500 mL
1 L
20 x 20 mL
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Pack
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
22365232 1 L Natural Poly Bottle
22365220 20 x 20 mL Pack
22365216 500 mL Natural Poly Bottle
2236521 4 L Natural Poly Bottle
2236525 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 22365232 Supplier Ricca Chemical Company Supplier No. R22365201A
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Conductivity Standard
Packaging Natural Poly Bottle
Name Note 1.413mS/cm (1413 μmho/cm) at 25°C
CAS 7647-14-5, 7732-18-5
CAS Min % 98.0, 0.07
CAS Max % 100.0, 0.07
Odor Odorless
Physical Form Liquid
Color Colorless
pH 6.7 to 7.3
Melting Point 0°C
Boiling Point 100°C
Solubility Completely miscible
Solution Type Conductivity Standard
Type Conductivity Standard
Quantity 1 L
Show More Show Less