Learn More
Conductivity Solution Standard, Hanna Instruments
Catalog No. 1160367
Encompass
Change view
Click to view available options
Quantity:
460 mL
Catalog No. Quantity
1160367 460 mL
1 options
Catalog No. 1160367 Supplier Hanna Instruments CDA Inc Supplier No. HI 70300L
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A storage solution prepared with reagent grade chemicals that can be used to ensure optimum performance of your pH and ORP electrodes.
  • Storage solution prepared with reagent grade chemicals that can be used to ensure optimum performance of your pH and ORP electrodes
  • Special formulation to minimize microbial growth and osmotic/diffusion effects between the solution and inner reference electrolyte
  • Manufactured specifically for pH and ORP electrodes
  • Clearly marked expiration date and lot number
Specifications
Chemical Name or Material Conductivity Solution Standard
Quantity 460 mL