Learn More
Conductivity Calibration Standards, YSI™
NIST-traceable conductivity solutions to calibrate both field instruments and lab instruments and cells together for maximum accuracy
Supplier:  Yellow Springs Instrument 060911
 View more versions of this product
Catalog No. 09-390-22
Only null left
Description
Description
  • Conductivity solutions are inspected with reference to primary standard solutions according to OIML recommendation 56
  • Bottles include a table of corrections at temperatures between 20 and 30°C
  • Available as: 1 Quart or in a box of 8 individual pints
Specifications
Specifications
Plastic Bottle
10,000μS/cm
Conductivity Calibration Standard
8 x 1 pt.
Product Suggestions
Product Suggestions
Videos
SDS Documents