missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ COMT Recombinant Rabbit Monoclonal Antibody (23GB3735)
GREENER_CHOICE

Catalog No. MA553456
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553456 100 μL
MA553457 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553456 Supplier Invitrogen™ Supplier No. MA553456
Only null left

Rabbit Recombinant Monoclonal Antibody

COMT Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

Catechol-O-methyltransferase catalyzes the transfer of a methyl group from S-adenosylmethionine to catecholamines, including the neurotransmitters dopamine, epinephrine, and norepinephrine. This O-methylation results in one of the major degradative pathways of the catecholamine transmitters. In addition to its role in the metabolism of endogenous substances, COMT is important in the metabolism of catechol drugs used in the treatment of hypertension, asthma, and Parkinson disease. COMT is found in two forms in tissues, a soluble form (S-COMT) and a membrane-bound form (MB-COMT). The differences between S-COMT and MB-COMT reside within the N-termini. Several transcript variants are formed through the use of alternative translation initiation sites and promoters.

Specifications

Antigen COMT
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3735
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Comt
Gene Accession No. P21964, P22734
Gene Alias Catechol O-methyltransferase; catecholamine-O-methyltransferase; catechol-O-methyltransferase; catechol-O-methyltransferase 1; catechol-O-methyltransferase isoform; Comt; Comt1; D16Wsu103e; D330014B15Rik; epididymis secretory sperm binding protein Li 98n; HEL-S-98n; OTTHUMP00000197749; OTTHUMP00000197750; testicular tissue protein Li 42
Gene Symbols Comt
Host Species Rabbit
Immunogen A synthetic peptide of human COMT (31-87AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1312, 24267
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less