missing translation for 'onlineSavingsMsg'
Learn More

Complex Nutrients - WP, MilliporeSigma™ Supelco™

This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.

Supplier:  Merck Emd Millipore PE10512ML

Catalog No. 111004917


Only null left
Explore available promotions

Description

Description

Featured Industries: Environmental
Specifications

Specifications

Concentrate for dilution to 1 L
Ampule
Proficiency Testing Material
2 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions