missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complete Volatiles Kit - WS, Proficiency Testing Sample, MilliporeSigma™ Supelco™
Description
Featured Industries: Environmental
Specifications
Specifications
| Packaging | Ampule |
| CAS | 67-56-1 |
| Flash Point | 9.7°C - closed cup |
| Purity Grade Notes | The PT sample is provided as a 1.2 mL concentrate.The solvent is methanol.The PT sample is not preserved. |
| Type | Proficiency Testing Material |
| Quantity | 1.2 mL |