missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek Complete Dispense System
Description
- Contains bottles, pumps, and tubing
Specifications
Specifications
| Type | Dispense System |
| For Use With (Equipment) | 406 FX Washer Dispenser |