missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Complement C4a Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5122178
Description
Complement C4a is a protein that is part of the complement system, which is a group of proteins that work together to help the immune system fight off infections. C4a is produced when the complement system is activated, and it plays a role in inflammation and the recruitment of immune cells to the site of infection. It is also involved in the clearance of immune complexes and the regulation of the complement system.Chemical Identifiers
| Complement C4a | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P0C0L4 | |
| 720 | |
| Asn680-Arg756, with N-terminal His and GST Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| C4a |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| C4A; C4AD; CO4A; Complement C4-A | |
| C4a | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| Antibody |
Specifications
| Complement C4a | |
| Polyclonal | |
| Unconjugated | |
| C4a | |
| C4A; C4AD; CO4A; Complement C4-A | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 720 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P0C0L4 | |
| C4a | |
| Asn680-Arg756, with N-terminal His and GST Tag. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |