Learn More
OHAUS™ Compass CR Series Scale
OHAUS™ Compass CR Series portable electronic scales are suitable for everyday weighing
Supplier: OHAUS™ 30428206
Description
- Intuitive two-button design makes for easy operation
- Slim, space-saving design allows for stackable storage
- Enlarged weighing platform can adequately handle large samples and a variety of objects
Specifications
| Product Safety: IEC/EN 61010-1; CAN/CSA C22.2 No. 61010-1; UL Std. No. 61010-1, Electromagnetic Compatibility: IEC/EN 61326-1 Class B, Basic environments; FCC Part 15 Class B; Canada ICES-003 Class B, Compliance Marks: CE; RCM | |
| LCD | |
| ABS housing and pan, slip-resistant feet, stability indicator, mechanical and software overload/underload protection, auto shut-off | |
| Portable Scale | |
| 8 in. | |
| 5.5 in. | |
| 1.6 in. | |
| 400 g |
| 2200 g | |
| 1 g | |
| 2 seconds | |
| 20.5 x 14 x 4.1 cm (8 x 5.5 x 1.6 in.) | |
| 205 mm | |
| 140 mm | |
| 41 mm |