missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Compact Safety Padlock Kit

Develop a safety plan to protect workers and the worksite using lockout products. The Brady™ Compact Safety Padlock Kit is a great solution for any worker needing just one lock and one tag. These kits are comprised of one compact safety padlock, one lockout tag, and a cable tie. Offered in nine different color formats.

$20.13

Specifications

Product Type Padlock Kit
Material Nylon
Dimensions (L x W) 0.125 x 1.2 in.
Includes (1) safety padlock; (1) lockout tag; (1) cable tie
Length (English) 0.125 in.
View More Specs

Products 1
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
19103595
View Documents Promotion Details
Brady™
123143
Red
Each for $20.13
Only null left
 
Description

Description

  • For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
  • Reduces the number of locks being used
  • Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
  • Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications

Specifications

Padlock Kit
0.125 x 1.2 in.
0.125 in.
1.2 in.
Nylon
(1) safety padlock; (1) lockout tag; (1) cable tie
1 Kit
Videos
SDS
Documents

Documents