Learn More
ABS Compact Pharmacy Refrigerator
Experience the best in vaccine refrigeration, certified to the NSF/ANSI 456 Vaccine Standard.
Supplier:  ABS PHABTNSF10PS
 View more versions of this product
Catalog No. 361058638
Only null left
Description
Description
These cutting-edge pharmacy refrigerators are certified in accordance with the NSF/ANSI 456 Standard for Vaccine Storage.
  • With this certification, units protect pharmaceuticals at optimal temperatures, preventing waste and allowing for peak delivery.
  • These compact glass door refrigerators utilize microprocessor controllers and feature temperature alarms, remote alarm contacts, and probe access ports with included probes.
  • Units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Specifications
Chemical Identifiers
Pharmacy Refrigerator
297 L
1
Scratch-resistant Painted Steel
Upright
H-drive
115 V
10.5 cu. ft.
1°C to 10°C
7
Powder-coated Steel
Microprocessor Temperature Controller
NEMA 5-15
Specifications
Pharmacy Refrigerator
297 L
Solid Swing
7
Powder-coated Steel
Upright
H-drive
115 V
10.5 cu. ft.
1°C to 10°C
1
Scratch-resistant Painted Steel
cULus
Microprocessor Temperature Controller
NEMA 5-15
Product Suggestions
Product Suggestions