missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Combs for Vertical Mini-Gels

Catalog No. 111011659
Change view
Click to view available options
No. of Wells:
1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Wells
111011659 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111011659 Supplier MilliporeSigma™ Supelco™ Supplier No. C26841EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Single Well, 1.5 mm

Specifications

Type Comb
For Use With Vertical Mini-Gels
No. of Wells 1
Width (Metric) 10 cm
Width (Metric) Well 1.5 mm