missing translation for 'onlineSavingsMsg'
Learn More

Combretastatin A4 98.0+%, TCI America™

Catalog No. C252025MG
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C252025MG 25 mg
C2520250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C252025MG Supplier TCI America Supplier No. C252025MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 117048-59-6
Molecular Formula C18H20O5
Molecular Weight (g/mol) 316.353
MDL Number MFCD03453309
InChI Key HVXBOLULGPECHP-WAYWQWQTSA-N
PubChem CID 5351344
IUPAC Name 2-methoxy-5-[(Z)-2-(3,4,5-trimethoxyphenyl)ethenyl]phenol
SMILES COC1=C(C=C(C=C1)C=CC2=CC(=C(C(=C2)OC)OC)OC)O

Specifications

Melting Point 83°C
Color White
Quantity 25 mg
Synonym (Z)-2-Methoxy-5-(3,4,5-trimethoxystyryl)phenol, 3′C-Hydroxy-3,4,4′C,5-tetramethoxy-cis-stilbene
Formula Weight 316.35
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Combretastatin A4
TSCA No
Recommended Storage Freezer