missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Column Frits

For Conventional Column Hardware (Compression Style)

Supplier:  MilliporeSigma™ Supelco™ SGMA59038

 View more versions of this product

Catalog No. 111011717


Only null left

Description

Description

  • Frits with 0.5 μm pores retain packings prepared from 3 μm particles
  • Frits with 2 μm pores retain larger particles
Specifications

Chemical Identifiers

Compression Style Column Hardware
Stainless Steel
Column Frit

Specifications

1/4 in.
Stainless Steel
Column Frit
Compression Style Column Hardware
0.5 μm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions