missing translation for 'onlineSavingsMsg'
Learn More

Color Standard, APHA/Hazen (Platinum-Cobalt), Color = 200, Ricca Chemical

Catalog No. 223020016
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
223020016 500 mL
R22302001C 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 223020016 Supplier Ricca Chemical Company Supplier No. 2230.20016
Only null left

Specifications

Chemical Name or Material Color Standard
Packaging Amber Glass Bottle
Name Note Color = 200
CAS Min % 95.0, 2.99, 0.03, 0.03
CAS Max % 99.0, 3.11, 0.03, 0.03
CAS 7647-01-0, 16921-30-5, 7791-13-1, 7647-01-0, 7732-18-5
Methods APHA / Hazen (Platinum-Cobalt)
Physical Form Liquid
Color Brown/Yellow to Yellow
Boiling Point 100°C
Quantity 500 mL
Show More Show Less