missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Collibri™ Stranded RNA Library Prep Kit for Illumina™ Systems

Catalog No. A38996024
Change view
Click to view available options
No. of Reactions:
24 Reactions
96 Reactions
Includes:
RNA Prep and UD Index
RNA Prep Kit Only
RNA Prep, Depletion Kit
RNA Prep, Depletion, UD Index
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions Includes
A38996024 24 Reactions RNA Prep and UD Index
A38994024 24 Reactions RNA Prep Kit Only
A38994096 96 Reactions RNA Prep Kit Only
A39003024 24 Reactions RNA Prep, Depletion Kit
A39003096 96 Reactions RNA Prep, Depletion Kit
A39005024 24 Reactions RNA Prep, Depletion, UD Index
A39005096 96 Reactions RNA Prep, Depletion, UD Index
A38996096 96 Reactions RNA Prep and UD Index
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. A38996024 Supplier Invitrogen™ Supplier No. A38996024
Only null left

Kits are designed for next-generation sequencing (NGS) on Illumina systems and support whole-transcriptome sequencing (human, mouse, rat), and mRNA sequencing (compatible with all species) applications.

The Invitrogen Collibri Stranded RNA Library Preparation kits are designed for next-generation sequencing (NGS) on Illumina systems. The kit supports whole-transcriptome sequencing (human, mouse, rat) and mRNA sequencing (compatible with all species) applications.

The Invitrogen Collibri Stranded RNA Library Preparation kits are designed for next-generation sequencing (NGS) on Illumina systems. The kit supports whole-transcriptome sequencing (human, mouse, rat) and mRNA sequencing (compatible with all species) applications.

The Collibri Stranded RNA Library Preparation Kit:
• Includes all necessary components in a single kit* for a quick use
• Total RNA converted to sequencing-ready library within 4.5 hours
• >98% strand specificity, efficient detection of non-coding RNA and high sensitivity of differential gene expression
• Provides in-process visual feedback for easier handling
• Is suitable for conventional automation to increase throughput

The Invitrogen Collibri Stranded RNA Library Prep Kit for Illumina Systems is designed for robust construction of cDNA libraries for strand-specific RNA sequencing on Illumina next-generation sequencing (NGS) systems. The kit brings together the superior features of SuperScript IV Reverse Transcriptase, Dynabeads magnetic particles, and Platinum SuperFi DNA Polymerase, enabling users to obtain high-quality sequencing-ready libraries. This Collibri Stranded RNA Library Prep Kit supports fast library preparation within 4.5 hours for mRNA or whole transcriptome libraries with 1–25 ng of mRNA-enriched or ribodepleted RNA sample input. Multiplexing of libraries can be carried out using up to 96 single-indexed or unique dual-indexed primers, enabling single-read or paired-end sequencing. The kit is suitable for various-quality RNA samples, including FFPE samples.

Visual cues of process integrity
For maximum convenience, visual cues of process integrity are included throughout the library preparation workflow. Monitor progress of library generation using visual feedback from dyes in critical reagents. The reaction mix changes color in response to the addition of critical components at each step to ensure that library preparation has the highest chance of success. The inert dyes do not interfere with enzymatic reactions and do not compromise sequencing results.

How it works
Full-length Illumina-compatible adaptors are introduced by PCR amplification using Platinum SuperFi DNA Polymerase to construct single-indexed or unique dual-indexed libraries compatible with single-read or paired-end sequencing. The kit contains 24 or 96 either single-indexed barcoded PCR primers (i7) pre-mixed with a universal i5 primer or unique dual-indexed PCR primers that enable multiplexing of up to 96 libraries. Optimized cleanup steps efficiently remove residual primers and adapter/primer dimers while preserving high library yields.

A related product, the Collibri Library Quantification Kit, is recommended for qPCR-based quantifications of libraries before proceeding to sequencing.

Applications
The Collibri Stranded RNA Library Prep Kit is recommended for:
• Gene expression studies
• Alternative splicing analysis
• Non-coding RNA detection and discovery
• Identification of alternative polyadenylation sites
• Gene fusion detection
• Detection of transcript isoforms

* The kit consists of enzymatic modification, size selection/clean-up, and indexed adapter-containing modules. Additional ribodepletion module of human/mouse/rat rRNA can also be included.

Specifications

Content And Storage • Collibri Library Prep Kit, store at -20°C
• Collibri Library Cleanup Kit, store at 4°C

Description The product contains modules A39117024, A39119024, A39100024
Sample Type RNA
For Use With (Equipment) Illumina NGS Instruments
Includes RNA Prep and UD Index
No. of Reactions 24 Reactions
Libraries cDNA Library
Product Line Collibri
Quantity 24 reactions
Sequencing Type Transcriptome Sequencing
Shipping Condition Approved for shipment on Wet or Dry Ice
Workflow Step Library Generation
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.