missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Collibri™ Stranded RNA Library Prep Kit for Illumina™ Systems

Catalog No. A38994024
Encompass
Change view
Click to view available options
Includes:
RNA Prep and UD Index
RNA Prep Kit Only
RNA Prep, Depletion Kit
RNA Prep, Depletion, UD Index
Index Included:
No
Yes
No. of Reactions:
24 Reactions
96 Reactions
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Index Included No. of Reactions
A38994024 RNA Prep Kit Only Yes 24 Reactions
A38994096 RNA Prep Kit Only Yes 96 Reactions
A39003024 RNA Prep, Depletion Kit No 24 Reactions
A39003096 RNA Prep, Depletion Kit No 96 Reactions
A39005024 RNA Prep, Depletion, UD Index Yes 24 Reactions
A39005096 RNA Prep, Depletion, UD Index Yes 96 Reactions
A38996024 RNA Prep and UD Index Yes 24 Reactions
A38996096 RNA Prep and UD Index Yes 96 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. A38994024 Supplier Invitrogen™ Supplier No. A38994024

Kits are designed for next-generation sequencing (NGS) on Illumina systems and support whole-transcriptome sequencing (human, mouse, rat), and mRNA sequencing (compatible with all species) applications.

The Invitrogen Collibri Stranded RNA Library Preparation kits are designed for next-generation sequencing (NGS) on Illumina systems. The kit supports whole-transcriptome sequencing (human, mouse, rat) and mRNA sequencing (compatible with all species) applications.

The Invitrogen Collibri Stranded RNA Library Preparation kits are designed for next-generation sequencing (NGS) on Illumina systems. The kit supports whole-transcriptome sequencing (human, mouse, rat) and mRNA sequencing (compatible with all species) applications.

The Collibri Stranded RNA Library Preparation Kit:

  • Includes all necessary components in a single kit* for a quick use
  • Total RNA converted to sequencing-ready library within 4.5 hours
  • >98% strand specificity, efficient detection of non-coding RNA and high sensitivity of differential gene expression
  • Provides in-process visual feedback for easier handling
  • Is suitable for conventional automation to increase throughput

The Invitrogen Collibri Stranded RNA Library Prep Kit for Illumina Systems is designed for robust construction of cDNA libraries for strand-specific RNA sequencing on Illumina next-generation sequencing (NGS) systems. The kit brings together the superior features of SuperScript IV Reverse Transcriptase, Dynabeads magnetic particles, and Platinum SuperFi DNA Polymerase, enabling users to obtain high-quality sequencing-ready libraries. This Collibri Stranded RNA Library Prep Kit supports fast library preparation within 4.5 hours for mRNA or whole transcriptome libraries with 1–25 ng of mRNA-enriched or ribodepleted RNA sample input. Multiplexing of libraries can be carried out using up to 96 single-indexed or unique dual-indexed primers, enabling single-read or paired-end sequencing. The kit is suitable for various-quality RNA samples, including FFPE samples.

Visual cues of process integrity

For maximum convenience, visual cues of process integrity are included throughout the library preparation workflow. Monitor progress of library generation using visual feedback from dyes in critical reagents. The reaction mix changes color in response to the addition of critical components at each step to ensure that library preparation has the highest chance of success. The inert dyes do not interfere with enzymatic reactions and do not compromise sequencing results.

How it works

Full-length Illumina-compatible adaptors are introduced by PCR amplification using Platinum SuperFi DNA Polymerase to construct single-indexed or unique dual-indexed libraries compatible with single-read or paired-end sequencing. The kit contains 24 or 96 either single-indexed barcoded PCR primers (i7) pre-mixed with a universal i5 primer or unique dual-indexed PCR primers that enable multiplexing of up to 96 libraries. Optimized cleanup steps efficiently remove residual primers and adapter/primer dimers while preserving high library yields.

A related product, the Collibri Library Quantification Kit, is recommended for qPCR-based quantifications of libraries before proceeding to sequencing.

Applications

The Collibri Stranded RNA Library Prep Kit is recommended for:

  • Gene expression studies
  • Alternative splicing analysis
  • Non-coding RNA detection and discovery
  • Identification of alternative polyadenylation sites
  • Gene fusion detection
  • Detection of transcript isoforms

* The kit consists of enzymatic modification, size selection/clean-up, and indexed adapter-containing modules. Additional ribodepletion module of human/mouse/rat rRNA can also be included.

Specifications

Content And Storage • Collibri Stranded RNA Library Prep Kit (24 preps). Store at -20°C.
• Collibri Library Cleanup Kit (24 preps). Store at 4°C.

Description The product contains modules A39117024, A39118024, A39100024
Format Kit
Species All
Sample Type mRNA
For Use With (Equipment) HiSeq X, HiSeq2500, HiSeq3000, HiSeq4000, MiSeq or iSeq, NextSeq 550, NextSeq 1000 and 2000, NovaSeq 6000
For Use With (Application) Sequencing
Includes RNA Prep Kit Only
No. of Reactions 24 Reactions
Libraries cDNA Libraries
Product Line Collibri, Illumina
Product Type Stranded RNA Library Prep Kit
Quantity 24 preps
Sequencing Type Transcriptome Sequencing
Shipping Condition Dry Ice
Workflow Step Adaptor Hybridization, Cleanup, Fragmentation, Indexing, Ligation, PCR Amplification, Reverse Transcription
Index Included Yes
Show More Show Less
Is the elution buffer, included in the Collibri Stranded RNA Library Prep Kit for Illumina Systems, "Tris buffered" or "Tris-Tween"?

Collibri Stranded RNA Library Prep Kit for Illumina Systems elution buffer is based on Tris-HCl, with no EDTA or EGTA.

Is there an RNA control for optimizing fragment length included with the Collibri Stranded RNA Library Prep Kit for Illumina Systems?

The Collibri Stranded RNA Library Prep Kit for Illumina Systems does not include an RNA control. Recommendations for RNA controls can be found in the Invitrogen Collibri Stranded RNA Library Prep Kit for Illumina User Guide, page 16 "Guidelines for RNA Controls".

What are the recommended RNA input sample types and respective sample amounts for Collibri Stranded RNA Library Prep Kit for Illumina Systems?

Information for RNA sample type and amount can be found in the Invitrogen Collibri Stranded RNA Library Prep Kit for Illumina User Guide, page 15 "Guidelines for RNA sample type and amount".

What are the differences between all Collibri Stranded RNA Library Prep Kit for Illumina Systems catalog numbers?

Information regarding the different catalog numbers can be found in the Invitrogen Collibri Stranded RNA Library Prep Kit for Illumina User Guide, page 3.
The different catalog numbers allow for the option to purchase different sizes of the library kit and with or without Unique Dual Indexes.

What additional materials are required when using Collibri Stranded RNA Library Prep Kit for Illumina Systems?

A list of required materials that are not supplied with the kit can be found in the Invitrogen Collibri Stranded RNA Library Prep Kit for Illumina User Guide, page 11-12.

What is the difference between the Collibri Stranded RNA Library Prep Kit for Illumina Systems Cat. Nos. A38994024 and A38996024?

Unlike Cat. No. A38994024, Cat. No. A38996024 comes with the Unique Dual Indexes (UDIs), listed in table 6, page 10 of the Invitrogen Collibri Stranded RNA Library Prep Kit for Illumina User Guide

Please also refer to the Technical Note "How to prevent index hopping" for additional information.

In general, UDIs have a sample index that is specific to a given library. All nucleic acids in a sample are labeled with the same sequence tag and the resulting library is pooled with other libraries and sequenced in parallel in a single run. UDIs are the preferred method when multiplexing to mitigate sample misassignment due to index hopping and the reagent kits supply enough to perform extra cycles. UDIs require extra cycles and the reagent kits supply enough to perform those extra cycles. UDIs can be especially effective when using instruments with patterned flow cells, such as the NovaSeq 6000 system.

Can indexed adaptors be used interchangeably between RNA-seq (RNA sequencing) and DNA-seq (DNA sequencing)?

No, indexed adaptors are specifically designed for each product.

Are adaptors included in the Collibri RNA library prep kits?

Yes. Ready-to-ligate adaptors are included in the Collibri RNA library prep kits. Please note that the adaptors are not available separately and are provided only as an integral part of the Collibri kits.

With the Collibri RNA library prep kits, when should the ERCC RNA Spike-In Control Mixes be added?

ERCC RNA Spike-In Control Mixes should be added to the sample before all other steps and should undergo exactly the same treatment as sample RNA. For whole transcriptome library generation, we recommend that you add ERCC RNA Spike-In Control Mixes to the input RNA before rRNA depletion or poly (A) selection.

With the Collibri RNA library prep kits, what is the advantage(s) of performing the cDNA step after the adaptor ligation?

Once the 5' and 3' adaptors are ligated to the RNA, cDNA synthesis starts not from random hexamers, but from one universal oligo that binds to the 3' adaptor. This eliminates bias from the priming site and also ensures conversion of every fragment into cDNA: random hexamers have to find a landing spot and some fragments do not have it and do not make it into the library. It also eliminates random nucleotide-introduced mistakes: during second-strand synthesis, a random hexamer becomes a legitimate part of the library, introducing “SNPs”, “point mutations” etc. These events are filtered out by software but nevertheless, decrease the output quality. With Collibri RNA library prep kit, adaptors never become part of the library.

Can the Collibri RNA library prep kits be adapted for automation?

Yes, the Collibri RNA library prep kits can be adapted for automation.


For Research Use Only. Not for use in diagnostic procedures.