missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ COLEC12 Monoclonal Antibody (K52057_2H12), Invitrogen™

Catalog No. MA556131
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556131 200 μL
MA555751 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556131 Supplier Invitrogen™ Supplier No. MA556131
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: VVEKMDNVTG GMETSRQTYD DKLTAVESDL KKLGDQTGKK AISTNSELST FRSDILDLRQ QLREITEKTS KNKDTLEKLQ ASGDALVDRQ SQLKETLENN SFLITTVNKT LQAYNGYVTN LQQDTSVLQG NLQNQMYSHN VVIMNLNNLN LTQVQQRNLI TNLQRSVDDT SQAIQRIKND FQNLQQVFLQ AKKDTDWLKE KVQSLQTLAA NNSALAKANN DTLEDMNSQL NSFTGQMENI TTISQANEQN LKDLQDLHKD AENRTAIKFN QLEERFQLFE TDIVNIISNI

This gene encodes a member of the C-lectin family, proteins that possess collagen-like sequences and carbohydrate recognition domains. This protein is a scavenger receptor, a cell surface glycoprotein that can bind to carbohydrate antigens on microorganisms facilitating their recognition and removal. In addition, these receptors can recognize oxidized phospholipids so they may also participate in removing oxidatively damaged or apoptotic cells.

Specifications

Antigen COLEC12
Applications Western Blot
Classification Monoclonal
Clone K52057_2H12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene Colec12
Gene Accession No. Q5KU26
Gene Alias CLP1; CL-P1; COLEC12; Collectin placenta protein 1; collectin subfamily member 12; collectin sub-family member 12; collectin-12; hCL-P1; NSR2; Nurse cell scavenger receptor 2; Scara4; Scavenger receptor class A member 4; scavenger receptor class A, member 4; scavenger receptor with C-type lectin; Srcl
Gene Symbols Colec12
Host Species Mouse
Immunogen Recombinant human COLEC12 (immunogen range is 61-350).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81035
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less