missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ShockWatch™ ColdMark™ Temperature Indicator
ShockWatch ColdMark Temperature Indicator is a single-use descending temperature indicator, is designed to help shippers identify and correct gaps in their cold chain.
$1,252.71
Specifications
| Certifications/Compliance | ROHS2, REACH |
|---|---|
| Disposable | Single-use |
| For Use With (Application) | Shipping and storage |
| Product Type | Disposable Temperature Indicator |
| Accuracy | ±1°C |
Alert:
This product contains no known hazardous or carcinogenic components at concentrations requiring reporting under OSHA 1200.
Description
- Turns from clear to violet when the temperature goes below a predetermined threshold
- Highly visible, easy to determine when cold chain has been broken
Order Info
Store above activation temperature and below 43°C / 110°F
Certifications
ROHS2, REACH
Warranty
2 years from date of sale
Specifications
| ROHS2, REACH | |
| Shipping and storage | |
| ±1°C |
| Single-use | |
| Disposable Temperature Indicator |