missing translation for 'onlineSavingsMsg'
Learn More

ShockWatch™ ColdMark™ Temperature Indicator

Catalog No. 22111032
Encompass
Change view
Click to view available options
Temperature Range (Metric):
2°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Range (Metric)
22111032 2°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This product contains no known hazardous or carcinogenic components at concentrations requiring reporting under OSHA 1200.
Catalog No. 22111032 Supplier ShockWatch™ Supplier No. CM 2/36

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This product contains no known hazardous or carcinogenic components at concentrations requiring reporting under OSHA 1200.

ShockWatch ColdMark Temperature Indicator is a single-use descending temperature indicator, is designed to help shippers identify and correct gaps in their cold chain.

  • Turns from clear to violet when the temperature goes below a predetermined threshold
  • Highly visible, easy to determine when cold chain has been broken

Order Info

Store above activation temperature and below 43°C / 110°F

Certifications

ROHS2, REACH

Warranty

2 years from date of sale

Specifications

Certifications/Compliance ROHS2, REACH
Disposable Single-use
For Use With (Application) Shipping and storage
Product Type Disposable Temperature Indicator
Max. Temperature (English) 36°F
Max. Temperature (Metric) 2°C
Min. Temperature (English) 36°F
Min. Temperature (Metric) 2°C
Temperature Range (English) 36°F
Temperature Range (Metric) 2°C
Accuracy ±1°C
Show More Show Less