Learn More
Fisherbrand™ Cold Water Faucet
Fisherbrand™ Cold Water Faucets are standard Laboratory cold water faucets.
 View more versions of this product
Catalog No. 06000397
Only null left
Description
Description
  • Deck mounted
  • Chrome plated brass with 8 in. swivel gooseneck
  • Valves are self-contained units with replaceable stainless steel seats
Specifications
Chemical Identifiers
Chrome-plated Brass
1 Ea.
Specifications
Chrome-plated Brass
1 Ea.
32.9 cm
30.5 cm
Blade Handle, Vacuum Breaker
13 in.
12 in.
Product Suggestions
Product Suggestions