missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Colchicine, 97%

Catalog No. p-3736030
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC227125000 500 mg Glass bottle
AC227120010 1 g Glass bottle
AC227120050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC227125000 Supplier Thermo Scientific Chemicals Supplier No. 227125000
Only null left

Colchicine, 7alphah-colchicine, CAS # 64-86-8, is a laboratory grade formulation of the anti-inflammatory plant alkaloid drug. | CAS: 64-86-8 | C22H25NO6 | 399.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Colchicine (7alphah-colchicine) is an alkaloid found in Colchicum autumnale that has been shown to have anti-gout and anti-inflammatory properties
  • Colchicine binds tubulin and prevents the migration of immune cells, reducing inflammation

Applications

  • Colchicine is an anti-mitotic; may be used in the study of cell proliferation and migration
  • Intended for in vivo and in vitro research applications
  • For research use only, not intended to treat disease

Chemical Identifiers

CAS 64-86-8
Molecular Formula C22H25NO6
Molecular Weight (g/mol) 399.44
MDL Number MFCD00078484,MFCD00078484
InChI Key IAKHMKGGTNLKSZ-INIZCTEOSA-N
Synonym colchicine, colchicina, colchicin, condylon, colchicinum, colchisol, colsaloid, colcin, colchineos, 7alphah-colchicine
PubChem CID 45038708
IUPAC Name 2,2,2-trideuterio-N-[(7S)-1,2,3-trimethoxy-9-oxo-10-(trideuteriomethoxy)-6,7-dihydro-5H-benzo[a]heptalen-7-yl]acetamide
SMILES COC1=C(OC)C(OC)=C2C(CC[C@H](NC(C)=O)C3=CC(=O)C(OC)=CC=C23)=C1

Specifications

Melting Point 150.0°C to 160.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. on anhydrous and solvent-free substance (HPLC)
Packaging Glass bottle
Quantity 500 mg
Beilstein 14,IV,946
Merck Index 15,2455
Specific Rotation − 250.00
Solubility Information Solubility in water: 45g/l (20°C). Other solubilities: soluble in alcohol and chloroform,practically insoluble in petroleum ether,1g/220mL ether,1g/100mL benzene
Formula Weight 399.44
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Colchicine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Fatal if swallowed.
May cause genetic defects.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-598-5

RUO – Research Use Only