missing translation for 'onlineSavingsMsg'
Learn More

Colchicine, 95% (dry wt.)

CAS: 64-86-8 | C22H25NO6 | 399.44 g/mol

Supplier:  Thermo Scientific Chemicals A1324014

 View more versions of this product

Catalog No. AAA1324014


Only null left
Explore available promotions

Description

Description

Colchicine is used as an anti-inflammatory agent for long-term treatment of behcet's Disease. It is used for constipation-predominant irritable bowel syndrome in women, and for treatment of severe or persistent aphthous stomatitis (canker sores).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water and alcohol. Sparingly soluble in ether.

Notes
Incompatible with strong oxidizing agents and mineral acids.
Specifications

Specifications

Colchicine
∼150°C (decomposition)
C22H25NO6
25 g
2228813
14,2471
Soluble in water and alcohol. Sparingly soluble in ether.
COC1=C(OC)C(OC)=C2C(CC[C@H](NC(C)=O)C3=CC(=O)C(OC)=CC=C23)=C1
399.44
399.44
64-86-8
(dry wt.)
MFCD00078484,MFCD00078484
UN1544
Light sensitive
colchicine, colchicina, colchicin, condylon, colchicinum, colchisol, colsaloid, colcin, colchineos, 7alphah-colchicine
IAKHMKGGTNLKSZ-INIZCTEOSA-N
N-[(10S)-3,4,5,14-tetramethoxy-13-oxotricyclo[9.5.0.0²,⁷]hexadeca-1(16),2,4,6,11,14-hexaen-10-yl]acetamide
45038708
95%

Chemical Identifiers

64-86-8
399.44
IAKHMKGGTNLKSZ-INIZCTEOSA-N
45038708
C22H25NO6
MFCD00078484,MFCD00078484
colchicine, colchicina, colchicin, condylon, colchicinum, colchisol, colsaloid, colcin, colchineos, 7alphah-colchicine
COC1=C(OC)C(OC)=C2C(CC[C@H](NC(C)=O)C3=CC(=O)C(OC)=CC=C23)=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H340
Fatal if swallowed.
May cause genetic defects.

P201-P202-P264b-P270-P281-P301+P310-P308+P313-P330-P501c

H300-H340

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ALKALOIDS, SOLID, N.O.S.

EINECSNumber : 200-598-5

RTECSNumber : GH0700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only