missing translation for 'onlineSavingsMsg'
Learn More

Colchicine, 95% (dry wt.)

Catalog No. AAA1324003
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1324003 1 g
AAA1324006 5 g
AAA1324014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1324003 Supplier Thermo Scientific Chemicals Supplier No. A1324003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 64-86-8 | C22H25NO6 | 399.44 g/mol

Colchicine is used as an anti-inflammatory agent for long-term treatment of behcet's Disease. It is used for constipation-predominant irritable bowel syndrome in women, and for treatment of severe or persistent aphthous stomatitis (canker sores).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water and alcohol. Sparingly soluble in ether.

Notes
Incompatible with strong oxidizing agents and mineral acids.

Chemical Identifiers

CAS 64-86-8
Molecular Formula C22H25NO6
Molecular Weight (g/mol) 399.44
MDL Number MFCD00078484,MFCD00078484
InChI Key IAKHMKGGTNLKSZ-INIZCTEOSA-N
Synonym colchicine, colchicina, colchicin, condylon, colchicinum, colchisol, colsaloid, colcin, colchineos, 7alphah-colchicine
PubChem CID 45038708
SMILES COC1=C(OC)C(OC)=C2C(CC[C@H](NC(C)=O)C3=CC(=O)C(OC)=CC=C23)=C1

Specifications

Melting Point ∼150°C (decomposition)
Quantity 1 g
UN Number UN1544
Beilstein 2228813
Sensitivity Light sensitive
Merck Index 14,2471
Solubility Information Soluble in water and alcohol. Sparingly soluble in ether.
IUPAC Name 2,2,2-trideuterio-N-[(7S)-1,2,3-trimethoxy-9-oxo-10-(trideuteriomethoxy)-6,7-dihydro-5H-benzo[a]heptalen-7-yl]acetamide
Formula Weight 399.44
Percent Purity 95%
Assay (dry wt.)
Chemical Name or Material Colchicine
Show More Show Less
Hazard Category H300-H340
Hazard Statement GHS H Statement
H300-H340
Fatal if swallowed.
May cause genetic defects.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P310-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ALKALOIDS, SOLID, N.O.S.
EINECSNumber 200-598-5
RTECSNumber GH0700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only