missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Colcemid, 98+%
Description
Microtubule modulator that depolymerizes microtubules and inactivates spindle fiber formation
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Quantity | 10 mg |
| Description | Pale yellow |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | Microtubule modulator that depolymerizes microtubules and inactivates spindle fiber formation |
| Formula Weight | 371.43 |
| Sensitivity | Air and Light Sensitive |
| Melting Point | 186°C |
| Solubility | Soluble in 10mg/ml chloroform. Also soluble in ethanol |
| Form | Powder |
| Beilstein | 2822892 |
| Show More |
Safety and Handling
| EINECSNumber | 207-514-6 |
| Recommended Storage | Keep cold |
RUO – Research Use Only