CAS RN 477-30-5
IUPAC Name:
3,4,5,14-tetramethoxy-10-(methylamino)tricyclo[9.5.0.0²,⁷]hexadeca-1(16),2,4,6,11,14-hexaen-13-one
Synonyms:
(+-)-demecolcine colchamine kolchicin demecolcin substance F kolchamin colcemide deacetyl-N-methylcolchicine methylcolchicine deacetylmethylcolchicine
Molecular Weight (g/mol):
371.43
Molecular Formula:
C21H25NO5
InChi Key:
NNJPGOLRFBJNIW-UHFFFAOYNA-N
SMILES:
CNC1CCC2=CC(OC)=C(OC)C(OC)=C2C2=CC=C(OC)C(=O)C=C12
1
–
1
of
1
results
1
Thermo Scientific Chemicals Colcemid, 98+%
CAS: 477-30-5 Molecular Formula: C21H25NO5 Synonym: Demecolcine; N-Methyl-N-desacetylcolchicine