missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Colcemid, 98+%

Catalog No. AAJ63900MF
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63900MF 50 mg
AAJ63900MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63900MF Supplier Thermo Scientific Chemicals Supplier No. J63900MF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 477-30-5 | C21H25NO5, C21H25NO5 | 371.43 g/mol

Microtubule modulator that depolymerizes microtubules and inactivates spindle fiber formation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 477-30-5
Molecular Formula C21H25NO5
Synonym Demecolcine; N-Methyl-N-desacetylcolchicine

Specifications

Quantity 50 mg
Description Pale yellow
Content And Storage 2°C to 8°C
For Use With (Application) Microtubule modulator that depolymerizes microtubules and inactivates spindle fiber formation
Formula Weight 371.43
Sensitivity Air and Light Sensitive
Melting Point 186°C
Solubility Soluble in 10mg/ml chloroform. Also soluble in ethanol
Form Powder
Assay 98+%

RUO – Research Use Only