missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ COL5A3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA583902

Catalog No. PIPA583902


Only null left

Description

Description

Immunogen sequence: STDIPKTETP APNLPPTPTP LVVTSTVTTG LNATILEGSL DPDSGTELGT LETKAAREDE EGDDSTMGPD FRAAEYPSRT QFQ Highest antigen sequence indentity to the following orthologs: Rat - 55%, Mouse - 52%.

Type I collagen, the fibrillar collagen found in most connective tissues, and the only component of the collagen found in cartilage. Mutations in this gene are associated with osteogenesis imperfecta, Ehlers-Danlos syndrome, and idiopathic osteoporosis. Reciprocal translocations between chromosomes 17 and 22, where this gene and the gene for platelet-derived growth factor beta are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans, resulting from unregulated expression of the growth factor. Two transcripts, resulting from the use of alternate polyadenylation signals, have been identified for this gene.
Specifications

Chemical Identifiers

COL5A3
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Polyclonal
Liquid
IgG
Human
COL5A3
COL5A3; Collagen 5; Collagen alpha-3(V) chain; collagen type V alpha 3; collagen, type V, alpha 3; collagen, type V, alpha 4; pro-(alpha)3(V) collagen; Pro-alpha3(V); procollagen, type V, alpha 3
COL5A3
Primary
Antigen affinity chromatography
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
RUO
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P25940
50509
Recombinant protein fragment of Human COL5A3 (Product #RP-98883)
Antibody

Specifications

COL5A3
Polyclonal
Unconjugated
COL5A3
COL5A3; Collagen 5; Collagen alpha-3(V) chain; collagen type V alpha 3; collagen, type V, alpha 3; collagen, type V, alpha 4; pro-(alpha)3(V) collagen; Pro-alpha3(V); procollagen, type V, alpha 3
Rabbit
Antigen affinity chromatography
RUO
50509
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P25940
COL5A3
Recombinant protein fragment of Human COL5A3 (Product #RP-98883)
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions