missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COL20A1 Polyclonal Antibody

Description
Antibody detects endogenous levels of total Collagen XX alpha 1.
Probable collagen protein.
Specifications
Specifications
| Antigen | COL20A1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | COL20A1 |
| Gene Accession No. | Q923P0, Q9P218 |
| Gene Alias | 1700051I12Rik; bA261N11.4; Col20a1; Collagen alpha-1(XX) chain; collagen type XX alpha 1 chain; collagen, type XX, alpha 1; collagen-like protein; KIAA1510; RGD1563901 |
| Gene Symbols | COL20A1 |
| Show More |