missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COL16A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149792
Description
COL16A1 Polyclonal Antibody for Western Blot, ICC/IF, IHC
This gene encodes the alpha chain of type XVI collagen, a member of the FACIT collagen family (fibril-associated collagens with interrupted helices). Members of this collagen family are found in association with fibril-forming collagens such as type I and II, and serve to maintain the integrity of the extracellular matrix. High levels of type XVI collagen have been found in fibroblasts and keratinocytes, and in smooth muscle and amnion.Specifications
| COL16A1 | |
| Polyclonal | |
| Unconjugated | |
| COL16A1 | |
| [a]1 (XVI) collagen; 2700007F12Rik; 447AA; A530052M23Rik; AI838662; alpha 1 type XVI collagen; CA1F; col16a1; Collagen alpha-1(XVI) chain; collagen type XVI alpha 1 chain; collagen XVI, alpha-1 polypeptide; collagen, type XVI, alpha 1; FP1572; procollagen type XVI alpha 1; procollagen, type XVI, alpha 1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 107581, 1307, 366474 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q07092, Q8BLX7 | |
| COL16A1 | |
| Recombinant Collagen Type XVI (COL16) (Val1434-Gly1604). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |