missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ COG8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529126
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529126 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529126 Supplier Invitrogen™ Supplier No. PA529126
Only null left

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, Raji. Predicted reactivity: Mouse (89%), Rat (91%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a protein that is a component of the conserved oligomeric Golgi (COG) complex, a multiprotein complex that plays a structural role in the Golgi apparatus, and is involved in intracellular membrane trafficking and glycoprotein modification. Mutations in this gene cause congenital disorder of glycosylation, type IIh, a disease that is characterized by under-glycosylated serum proteins, and whose symptoms include severe psychomotor retardation, failure to thrive, seizures, and dairy and wheat product intolerance.

Specifications

Antigen COG8
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene COG8
Gene Accession No. Q96MW5
Gene Alias BB235941; C87832; CDG2H; COG complex subunit 8; Cog8; Component of oligomeric Golgi complex 8; conserved oligomeric golgi complex component 8; conserved oligomeric Golgi complex protein 8; conserved oligomeric Golgi complex subunit 8; dependent on RIC1; DOR1; MNCb-5704
Gene Symbols COG8
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human COG8. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84342
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less