missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COG7 Polyclonal Antibody

Description
Recommended positive controls: HeLa mf. Predicted reactivity: Mouse (97%), Rat (96%), Zebrafish (81%), Xenopus laevis (84%), Chicken (82%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a WD repeat-containing protein that interacts with the Cul4-Ddb1 E3 ligase macromolecular complex. Multiple alternatively spliced transcript variants have been found for this gene.
Specifications
Specifications
| Antigen | COG7 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | COG7 |
| Gene Accession No. | P83436 |
| Gene Alias | 5630400E24Rik; CDG2E; COG complex subunit 7; Cog7; Component of oligomeric Golgi complex 7; conserved oligomeric Golgi complex subunit 7; Gm167; UNQ3082/PRO10013 |
| Gene Symbols | COG7 |
| Show More |