missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ COG3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584200
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584200 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584200 Supplier Invitrogen™ Supplier No. PA584200

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

Immunogen sequence: DGQLFLIKHL LILREQIAPF HTEFTIKEIS LDLKKTRDAA FKILNPMTVP RFFRLNSNNA LIEFLLEGTP EIREHYLDSK KDVDRHLKSA CEQFIQQQT Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.

This gene encodes a component of the conserved oligomeric Golgi (COG) complex which is composed of eight different subunits and is required for normal Golgi morphology and localization. Defects in the COG complex result in multiple deficiencies in protein glycosylation. The protein encoded by this gene is involved in ER-Golgi transport.

Specifications

Antigen COG3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COG3
Gene Accession No. Q96JB2
Gene Alias COG complex subunit 3; COG3; component of oligomeric golgi complex 2; component of oligomeric golgi complex 3; Conserved oligomeric Golgi complex subunit 3; E430004N23; p94; SEC34; tethering factor SEC34; vesicle-docking protein SEC34 homolog
Gene Symbols COG3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COG3. Recombinant protein control fragment (Product #RP-108403).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83548
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less