missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cofilin Monoclonal Antibody (C12), Invitrogen™
GREENER_CHOICE

Catalog No. FEMA551063
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551063 200 μL
FEMA551064 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEMA551063 Supplier Invitrogen™ Supplier No. MA551063
Only null left

Mouse Monoclonal Antibody

Cofilin binds to F-actin and exhibits pH-sensitive F-actin depolymerizing activity. Regulates actin cytoskeleton dynamics. Important for normal progress through mitosis and normal cytokinesis. Plays a role in the regulation of cell morphology and cytoskeletal organization. Required for the up-regulation of atypical chemokine receptor ACKR2 from endosomal compartment to cell membrane, increasing its efficiency in chemokine uptake and degradation. Required for neural tube morphogenesis and neural crest cell migration.

Specifications

Antigen Cofilin
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone C12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4
Gene CFL1
Gene Accession No. P10668, P23528, Q5E9F7
Gene Alias 18 kDa phosphoprotein; AA959946; CFL; Cfl1; CFL-1; Cof; COF1; cofilin; cofilin 1; cofilin 1 (non-muscle); cofilin 1, non-muscle; cofilin, non-muscle isoform; Cofilin1; Cofilin-1; cofilin-1-like protein; epididymis secretory protein Li 15; HEL-S-15; n-cofilin; p18
Gene Symbols CFL1
Host Species Mouse
Immunogen Recombinant Human CFL1 protein, Ser3-Leu161 (Accession #P23528).
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1072, 445532, 534553
Target Species Bovine, Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less