missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Coenzyme B12

Catalog No. AC464362500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464362500 250 mg
AC464360010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464362500 Supplier Thermo Scientific Chemicals Supplier No. 464362500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13870-90-1 | C72H100CoN18O17P | 1579.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13870-90-1
Molecular Formula C72H100CoN18O17P
Molecular Weight (g/mol) 1579.61
InChI Key OAJLVMGLJZXSGX-UHFFFAOYNA-L
IUPAC Name cobalt(3+) 4,9,14-tris(2-carbamoylethyl)-3,8,19-tris(carbamoylmethyl)-18-{2-[(2-{[5-(5,6-dimethyl-1H-1,3-benzodiazol-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl phosphonato]oxy}propyl)carbamoyl]ethyl}-2,3,6,8,13,13,16,18-octamethyl-20,21,22,23-tetraazapentacyclo[15.2.1.1²,⁵.1⁷,¹⁰.1¹²,¹⁵]tricosa-5(23),6,10(22),11,15(21),16-hexaen-20-ide [5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methanide
SMILES [Co+3].[CH2-]C1OC(C(O)C1O)N1C=NC2=C(N)N=CN=C12.CC(CNC(=O)CCC1(C)C(CC(N)=O)C2[N-]C1=C(C)C1=NC(=CC3=NC(=C(C)C4=NC2(C)C(C)(CC(N)=O)C4CCC(N)=O)C(C)(CC(N)=O)C3CCC(N)=O)C(C)(C)C1CCC(N)=O)OP([O-])(=O)OC1C(CO)OC(C1O)N1C=NC2=CC(C)=C(C)C=C12

Specifications

Quantity 250 mg
Formula Weight 1579.58
Chemical Name or Material Coenzyme B12
EINECSNumber 237-627-6

RUO – Research Use Only