missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Coelenterazine h

Catalog No. C6780
Encompass
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C6780 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C6780 Supplier Molecular Probes™ Supplier No. C6780

Coelenterazine h can be used to reconstitute the aequorin complex both in vivo and in vitro.

Coelenterazine h is one of several synthetic coelenterazine analogs that confer different Ca2+ affinities and spectral properties on the aequorin complex. Like native coelenterazine, these five novel derivatives can be used to reconstitute the aequorin complex both in vivo and in vitro.

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Bioluminescence
For Use With (Equipment) Luminometer (Microplate), Luminometer (Tube-based)
Product Type Coelenterazine
Dye Type Protein-Based
Quantity 250 μg
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.