missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COD Standard, 1000ppm (1mL = 1mg COD) (Potassium Acid Phthalate in Water), Certified, LabChem™
Specifications
Specifications
| Packaging | Poly Bottle |
| CAS Max % | 99.61, 0.09, 0.3 |
| Identification | Passes Test |
| Physical Form | Liquid |
| Concentration | 1000 ppm |
| Linear Formula | KHC8H4O4 |
| Color | Colorless |
| Solubility | Soluble in water |
| Quantity | 500 mL |
| Formula Weight | 204.22 |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement Solution is not hazardous. |
| Precautionary Statement | GHS P Statement If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes. If swallowed, get medical attention. |
| Recommended Storage | Room Temperature |