missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cochineal Red A, Spectrum™ Chemical
$205.61 - $26103.93
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18609880
|
Spectrum Chemical Mfg Cor
CO10225GM |
25 g | Amber Glass Bottle |
Each for $205.61
|
|
||||
|
18609879
|
Spectrum Chemical Mfg Cor
CO102100GM |
100 g | Amber Glass Bottle |
Each for $508.86
|
|
||||
|
18609881
|
Spectrum Chemical Mfg Cor
CO10250KGBL |
50 kg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 2611-82-7 |
Specifications
| 2611-82-7 | |
| Reagent | |
| Maroon | |
| Cochineal Red A |
| 1 | |
| Amber Glass Bottle | |
| 25 g |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.