missing translation for 'onlineSavingsMsg'
Learn More

Cochineal Red A, Spectrum™ Chemical

Catalog No. 18609880
Change view
Click to view available options
Quantity:
25 g
100 g
50 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18609880 25 g Amber Glass Bottle
18609879 100 g Amber Glass Bottle
18609881 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18609880 Supplier Spectrum Chemical Mfg Cor Supplier No. CO10225GM
Only null left

Chemical Identifiers

CAS 2611-82-7

Specifications

CAS Max % 1
Grade Reagent
Packaging Amber Glass Bottle
Color Maroon
Quantity 25 g
Chemical Name or Material Cochineal Red A

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.